missing translation for 'onlineSavingsMsg'
Learn More

ORP Buffer 7 (87 mV), Reagents

Catalog No. 01334570
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334570 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334570 Supplier Labchem Inc Supplier No. CO0052001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ORP Buffer 7 (87 mV), Reagent, use for Redox. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material ORP Buffer 7 (87 mV)
Packaging Poly Bottle
CAS 7732-18-5, 7558-79-4, 7778-77-0
Specific Gravity 1.007
Physical Form Liquid
Technique Redox
Color Colorless
pH 7
Boiling Point 100°C
For Use With (Equipment) Redox Meter
Synonym Oxidation Reduction Potential Buffer
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Show More Show Less