missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Ornidazole | |
| 92°C | |
| C7H10ClN3O3 | |
| 25 g | |
| IPWKIXLWTCNBKN-UHFFFAOYSA-N | |
| 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol | |
| 28061 | |
| 219.63 | |
| Crystalline Powder |
| 16773-42-5 | |
| White-Yellow | |
| MFCD00057960 | |
| ornidazole, madelen, tiberal, ornidazolum, 1-3-chloro-2-hydroxypropyl-2-methyl-5-nitroimidazole, ornidazol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl propan-2-ol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl-2-propanol, ornidal, 1-2-hydroxy-3-chloropropyl-2-methyl-5-nitroimidazole | |
| CC1=NC=C(N1CC(CCl)O)[N+](=O)[O-] | |
| 219.625 | |
| CHEBI:75176 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 16773-42-5 | |
| 219.625 | |
| IPWKIXLWTCNBKN-UHFFFAOYSA-N | |
| 28061 | |
| 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol |
| C7H10ClN3O3 | |
| MFCD00057960 | |
| ornidazole, madelen, tiberal, ornidazolum, 1-3-chloro-2-hydroxypropyl-2-methyl-5-nitroimidazole, ornidazol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl propan-2-ol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl-2-propanol, ornidal, 1-2-hydroxy-3-chloropropyl-2-methyl-5-nitroimidazole | |
| CHEBI:75176 | |
| CC1=NC=C(N1CC(CCl)O)[N+](=O)[O-] |
Safety and Handling
RTECSNumber : NI5460000
TSCA : No