missing translation for 'onlineSavingsMsg'
Learn More

Ornidazole 98.0+%, TCI America™

Catalog No. p-7134325
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04175G 5 g
O041725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04175G Supplier TCI America Supplier No. O04175G
Only null left

Chemical Identifiers

CAS 16773-42-5
Molecular Formula C7H10ClN3O3
Molecular Weight (g/mol) 219.625
MDL Number MFCD00057960
InChI Key IPWKIXLWTCNBKN-UHFFFAOYSA-N
Synonym ornidazole, madelen, tiberal, ornidazolum, 1-3-chloro-2-hydroxypropyl-2-methyl-5-nitroimidazole, ornidazol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl propan-2-ol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl-2-propanol, ornidal, 1-2-hydroxy-3-chloropropyl-2-methyl-5-nitroimidazole
PubChem CID 28061
ChEBI CHEBI:75176
IUPAC Name 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
SMILES CC1=NC=C(N1CC(CCl)O)[N+](=O)[O-]

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ornidazole
RTECSNumber NI5460000
TSCA No