missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Versa Star Pro™ pH/ISE Benchtop Multiparameter Meter

Get premium performance and ultimate flexibility with our benchtop meter with pH/ISE module for pH, ion concentration, and temperature.

Supplier:  Thermo Scientific™ VSTAR40B

 View more versions of this product

Catalog No. 13645581


Only null left

Description

Description

Get high accuracy, premium performance and ultimate flexibility with the Thermo Scientific™ Orion™ Versa Star Pro™ pH/ISE Benchtop Multiparameter Meter. Meet your most challenging applications for pH, ion concentration, mV, ORP and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. Perform up to 6 point pH and ISE calibrations, log up to 2000 data point sets with time/date stamp and transfer calibration and data logs via USB or RS232 to a printer or computer. Select systems with one or two pH/ISE modules.

Thermo Scientific Orion Versa Star Pro Benchtop Meters are designed to meet your most challenging application with ease. Delivering rich functionality and features, you can optimally configure your meter system to meet changing requirements. The Orion Versa Star Pro Benchtop Meters offer interchangeable measurement modules that allow multiple users to customize four separate channels to meet their specific requirements. Easily view your measurements on a stunning color display with adjustable text sizes. Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages. Enjoy quick and simple data entry using the numeric keypad.

  • Onscreen measurement stability indicator lets you know exactly when your measurements are ready
  • Selectable read modes include Auto-Read to hold a stable reading, Continuous to show changing readings, Timed to log data at specific intervals and Single-Shot to hold a reading after a specific wait time is reached
  • Audible alarms automatically notify you when a set offset value, high/low limit value and calibration time interval are reached
  • Easily access, update and switch your preferred measurement settings, using 10 password-protected methods per channel
  • Uniformly mix solutions using up to two meter controlled stirrer probes that are easy to position and can be quickly rinsed between samples
  • Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID and account ID
  • Calibration log stores 30 most recent calibrations per parameter with viewable graph for multipoint pH and ISE calibrations
  • Ensure measurement integrity by viewing the active calibration log while taking readings
  • Non-volatile meter memory preserves data and settings, even in the event of power loss
  • Meter facilitates data transfers to a computer via USB or RS232 connections
  • IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
  • Meters include CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty

Includes Orion Versa Star pH/ISE Measurement Module:

  • Measure pH, ion concentration, mV, relative mV or ORP with temperature
  • Perform up to a 6 point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Perform up to a 6 point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb or none
  • Incremental techniques include single known addition, double known addition, single known subtraction, double known subtraction, analate addition and analate subtraction
  • Advanced ISE features include non-linear selectable auto-blank, low range stability and electrode ID with calibration verification
  • Select the VSTAR40B meter or VSTAR40B2 kit with two pH/ISE modules to measure multiple samples simultaneously

System Access Feature
Orion Versa Star Pro meters offer a system access feature that is designed to help ensure data integrity by preventing undesired access to various meter functions including date/time setting, data log and calibration log. The System Access feature can be easily turned on or off as the needs of your lab change for ultimate flexibility in operation and ease-of-use. When enabled, the System Access feature allows assignment of a system administrator and up to 10 user accounts with customizable access to instrument settings, measurement settings, calibrations and logs. Use the password protection to secure selected meter functions, while leaving other functions accessible for all users. The System Access feature allows you to customize meter access to meet your specific security and data reporting needs.

Specifications

Chemical Identifiers

USB (2), RS232
±0.2 mV or ±0.05% of reading whichever is greater
±0.002
Set point alarm, high/low limit alarm, calibration due alarm
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
1 to 6
CE, TUV 3-1, FCC Class A
2000 with Date and Time Stamp
Color graphic LCD with user-customizable options
pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature
-2 to 20
0.1/0.01/0.001
7.000pH (adjustable), 1.000 ISE (adjustable)
English, Spanish, French, Italian, German, Chinese, Portuguese and Korean
Non-volatile memory preserves data log, calibration log and meter settings
DC input: 9VDC, 1A
0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none
±2000.0 mV
0.1mV
±0.1°C
5°C to 45°C (ambient)
22.0°F to 221.0° F
Manual or automatic with ATC temperature probe
28cm x 24cm x 10cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H
1.2 kg, modules add 0.2 kg each
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR)
±0.2 mV or ±0.05% of reading whichever is greater
Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit
Yes
1 point relative mV (RmV) mode, 1 point ORP mode
1 to 6
Transfer single, range or all data points to printer or computer
4 (Customizable with modules)
IP54
±2000.0 mV
0.0001 minimum, 1 to 4 significant digits (user selectable)
-5.0°C to +105.0°C
Numeric with menu-specific function keys and dual purpose scroll/shortcut keys
Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with
10 per channel
Universal AC adapter (included)
±2000.0 mV
5 to 85 %, non-condensing
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
41°F to 113°F (ambient)
1 point temperature offset calibration
0.1°C, 0.1°F
3 year meter and module warranty
2.5 lb., modules add 0.5 lb. each

Specifications

Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR)
±0.2 mV or ±0.05% of reading whichever is greater
±0.002
Set point alarm, high/low limit alarm, calibration due alarm
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
1 to 6
CE, TUV 3-1, FCC Class A
2000 with Date and Time Stamp
4 (Customizable with modules)
IP54
±2000.0 mV
0.0001 minimum, 1 to 4 significant digits (user selectable)
-5.0°C to +105.0°C
Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe)
Numeric with menu-specific function keys and dual purpose scroll/shortcut keys
Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with
10 per channel
Universal AC adapter (included)
Meter, 2 Channel pH/Ion (Sensor Not Included)
±2000.0 mV
5 to 85 %, non-condensing
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
41°F to 113°F (ambient)
1 point temperature offset calibration
0.1°C, 0.1°F
3 year meter and module warranty
2.5 lb., modules add 0.5 lb. each
USB (2), RS232
±0.2 mV or ±0.05% of reading whichever is greater
Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit
Yes
1 point relative mV (RmV) mode, 1 point ORP mode
1 to 6
Transfer single, range or all data points to printer or computer
Orion Versa Star Pro benchtop meter with two pH/ISE/mV modules and stand
Color graphic LCD with user-customizable options
pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature
-2 to 20
0.1/0.01/0.001
Versa Star Pro meter, two VSTAR-ISE pH/ISE modules, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate
7.000pH (adjustable), 1.000 ISE (adjustable)
English, Spanish, French, Italian, German, Chinese, Portuguese and Korean
Non-volatile memory preserves data log, calibration log and meter settings
DC input: 9VDC, 1A
pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (purchase separately)
0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none
±2000.0 mV
0.1mV
±0.1°C
5°C to 45°C (ambient)
22.0°F to 221.0° F
Manual or automatic with ATC temperature probe
28cm x 24cm x 10cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H
1.2 kg, modules add 0.2 kg each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions