missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion Star™ A325 pH/Conductivity Portable Multiparameter Meter

Catalog No. 13645565
Change view
Click to view available options
Product Type:
Portable Meter
Portable Meter Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
13645565 Portable Meter Kit
13645563 Portable Meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13645565 Supplier Thermo Scientific™ Supplier No. STARA3255

Accurately measure pH, conductivity and temperature with our pH/conductivity portable meter for high performance in challenging field tests.

Get high accuracy and premium performance everywhere you need it with the Thermo Scientific™ Orion Star™ A325 pH/Conductivity Portable Multiparameter Meter. This meter is designed for a wide range of pH, mV, ORP, conductivity, total dissolved solids (TDS), salinity, resistivity and temperature testing and field applications. Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Use the two channels to measure pH and conductivity simultaneously or view each channel separately. Log up to 5000 data point sets with time/date stamp and transfer logs to a printer or computer.

Take your measurements at the source with our premium line of portable meters. The Thermo Scientific Orion Star A325 pH/Conductivity Portable Meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the elements with you. Orion Star A325 pH/Conductivity Portable Meters are designed to be easy-to-use, dependable field meters that offer advanced features and functions including a 5000 point date log with sample ID and user ID options, calibration and high/low limit alarms and password-protected measurement methods.

  • Combined convenience with the benefits of a pH meter and conductivity meter in one easy-to-use system
  • Measure pH, mV, relative mV or ORP with temperature on channel one and measure conductivity, TDS, salinity or resistivity with temperature on channel two
  • Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously
  • Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Perform up to a 5 point conductivity calibration
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
  • Use linear, non-linear, EP (USP) or off conductivity temperature compensation with 5°C, 10°C, 15°C, 20°C or 25°C reference temperatures
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
  • Calibration log stores 10 most recent calibrations
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty

Specifications

AC Adapter Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Outputs USB, RS232
Accuracy (Conductivity) 0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
Accuracy (Resistivity) 0.5% of reading ±1 digit
Accuracy (Salinity) 0.5% of reading ±1 digit
Accuracy (TDS) 0.5% of reading ±1 digit
Alarm Output High/low limit alarm, calibration due alarm
Backlight Option Yes (Selectable)
Battery Life 800 hr.
Battery Type 4 AA (included)
Calibration pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op
Calibration Points 1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (Conductivity) Up to 5
Calibration Points (pH) 1 to 5
Cell Constant 0.001 to 199.9cm-1
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Management Details Transfer single, range or all data points to printer or computer
Data Points 5000 with Date and Time Stamp
Description Orion Star A325 pH/conductivity portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, DuraProbe 4-cell conductivity sensor, solutions, meter armor and field case
Channels 2
Display Type LCD Graphic, Backlit
IP Rating IP67
Measurement Modes pH, mV, Relative mV, TDS, ORP with Temperature, Salinity, Conductivity, Resistivity with Temperature
Range (Conductivity) 0.001 μS/cm to 3000 mS/cm
Range (mV) ±2000.0 mV
Range (pH) -2 to 20
Range (Resistivity) 2 ohm to 100.0 meg-ohm
Range (Salinity) 0.06 to 80.00 psu, 0.05 to 42.00 ppt
Range (TDS) 0.001 ppm to 200.0 ppt
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) -5.0°C to +105.0°C
Includes Orion STAR A325 pH/conductivity portable meter; 8107UWMMD Orion ROSS Triode pH/ATC gel-filled electrode with 3m cable; 013010MD Orion DuraProbe conductivity cell with 3m cable; 910410 pH 4.01 buffer pouches, pack of 10; 910710 pH 7.00 buffer pouches, pack of 10; 911010 pH 10.01 buffer pouches, pack of 10; 01100710 conductivity standard pouches, 1413 uS; 911110 rinse solution, pack of 10; protective armor with electrode holder; hard carrying case and batteries
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe)
Isopotential Point 7.000pH
Keypad Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Log Function Type Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
Memory Non-volatile memory preserves data log, calibration log and meter settings
Methods 10 per channel with password protection
Model Orion Star A325 pH/mV/RmV/ORP/Conductivity/TDS/Salinity/Resistivity/Temperature Portable Meter
Power Supply 4 AA batteries (included) or universal AC adapter (sold separately)
Product Type Portable Meter Kit
Range (ORP) ±2000.0 mV
Range (Relative mV) ±2000.0 mV
Relative Humidity Range 5 to 85 %, non-condensing
Resolution (Conductivity) 0.001 μS minimum, auto ranging up to 4 significant digits
Resolution (mV) 0.1mV
Resolution (Resistivity) 1 ohm or 0.1 meg-ohm, auto ranging
Resolution (Salinity) 0.01 psu or 0.01 ppt minimum, auto ranging
Resolution (TDS) 0.001 ppm minimum, auto ranging up to 4 significant digits
Salinity Measurement Modes Practical salinity (psu) or natural sea water (ppt)
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
TDS Factor Range Linear (0.02 to 9.99)
Temperature Accuracy ±0.1°C
Temperature Compensation Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature (Metric) Reference 5, 10, 15, 20, 25°C
Temperature Probe Calibration 1 point temperature offset calibration
Temperature Range (English) 22.0°F to 221.0° F
Temperature Resolution 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Warranty 3 Year Meter
Dimensions (L x W x H) 24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.)
Weight (English) 1.2 lb.
Weight (Metric) 0.5 kg
Show More Show Less