Learn More
Thermo Scientific™ Orion Star™ A322 Conductivity Portable Meter
Accurately monitor conductivity, TDS, salinity, resistivity and temperature with this conductivity portable meter designed for challenging field tests.
$2,609.69 - $3,849.74
Specifications
Accuracy (Conductivity) 0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Outputs USB, RS232
Measurement Modes Conductivity, TDS, Salinity, Resistivity with Temperature
Compatible Probes 2-cell or 4-cell conductivity sensors with built-in temperature
Cell Constant 0.001 to 199.9cm-1
View More Specs
Products 3
Catalog Number Mfr. No. Probe Type Price Quantity & Availability  
Catalog Number Mfr. No. Probe Type Price Quantity & Availability  
13645554
View Documents Promotion Details
Thermo Scientific™
STARA3220
Not included
Each for $2,609.69
Only null left
13303213
View Documents Promotion Details
Thermo Scientific™
STARA3226
2-cell Conductivity Sensor
Each for $3,849.74
Only null left
13645556
View Documents Promotion Details
Thermo Scientific™
STARA3225
4-cell Conductivity Sensor
Each for $3,720.19
Only null left
Description
Description
Get high accuracy and premium performance everywhere you need it with the Thermo Scientific™ Orion Star™ A322 Conductivity Portable Meter. This instrument is designed for a wide range of conductivity, total dissolved solids (TDS), salinity, resistivity and temperature testing and field applications. Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Select linear, non-linear pure water or EP temperature compensation with 5°C, 10°C, 15°C, 20°C or 25°C reference temperatures. Log up to 5000 data point sets with time/date stamp and transfer to a printer or computer.
Take your measurements at the source with our premium line of portable meters. The Thermo Scientific Orion Star A322 conductivity portable meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the elements with you. Orion Star A322 conductivity portable meters are designed to be easy-to-use, dependable field meters that offer advanced features and functions including a 5000 point date log with sample ID and user ID options, calibration and high/low limit alarms and password-protected measurement methods.
  • Measure conductivity, TDS, salinity or resistivity with temperature
  • Perform up to a five point conductivity calibration with calibration editing option that allows individual points to be fixed without a full recalibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
  • Use linear, non-linear, EP (USP) or off conductivity temperature compensation with 5°C, 10°C, 15°C, 20°C or 25°C reference temperatures
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
  • Calibration log stores ten most recent calibrations
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications and 3-year replacement warranty
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Conductivity, TDS, Salinity, Resistivity with Temperature
0.001 to 199.9cm-1
5
0.001 μS minimum, auto ranging up to 4 significant digits
5, 10, 15, 20, 25°C
0.01 psu or 0.01 ppt minimum, auto ranging
Practical salinity (psu) or natural sea water (ppt)
0.5% of reading ±1 digit
-5°C to +105°C
±0.1°C
Manual or automatic with ATC temperature probe
Graphic LCD with Backlight
Non-volatile memory preserves data log, calibration log and meter settings
Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
High/low limit alarm, calibration due alarm
CE, TUV 3-1, FCC Class A
24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
1.2 lb.
Orion Star A322 Conductivity/TDS/Salinity/Resistivity/Temperature Portable Meter
4 AA batteries (included) or universal AC adapter (sold separately)
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
5°C to 45°C (ambient)
0.5% of reading ±1 digit
800 hr.
USB, RS232
2-cell or 4-cell conductivity sensors with built-in temperature
Conductivity with calibration editing option and temperature
0.001 μS/cm to 3000 mS/cm
Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off wit
0.06 to 80.00 psu, 0.05 to 42.00 ppt
0.5% of reading ±1 digit
0.001 ppm minimum, auto ranging up to 4 significant digits
Linear (0.02 to 9.99)
0.1°C (0.1°F)
2 ohm to 100.0 megohm
1 point temperature offset calibration
Yes (Selectable)
5000 with Date and Time Stamp
Transfer single, range or all data points to printer or computer
8 pin MiniDIN (conductivity sensor with built-in temperature)
IP67
0.5 kg
5 to 85%, non-condensing
1 ohm or 0.1 megohm, auto ranging
10 per channel with password protection
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
41°F to 113°F (ambient)
1
4 x AA (included)