Learn More
Thermo Scientific™ Orion™ All-in-One™ pH Buffer Kits
Description
Thermo Scientific™ Orion™ All-in-One™ pH buffer kits include pH 4.01, 7.00 and 10.01 buffers that are color-coded for easy identification as well as storage solution to help ensure your pH electrode is properly maintained. Using high-quality pH buffers to calibrate your pH electrode and pH meter is the best way to ensure your pH measurements are accurate and reproducible. Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.
Kit includes one each:
- 1 pint (475mL) bottle of pH 4.01 buffer (910104)
- 1 pint (475mL) bottle of pH 7.00 buffer (910107)
- 1 pint (475mL) bottle of pH 10.01 buffer (910110)
- 1 pint (475mL) bottle of ROSS™ pH electrode storage solution (810001)
- 1 oz. (60mL) bottle of general pH electrode cleaning solution (900024)
- 12mm diameter pH electrode storage bottle
Specifications
Specifications
| Description | ROSS All-in-One pH Buffer Kit |
| For Use With (Application) | Accurate calibrations and NIST traceability, laboratory and bench analysis |
| pH | 4.01, 7.00, 10.01 |
| Quantity | 4 x 475 mL, 1 x 60 mL |
| Solution Type | pH Buffer, Storage Solution and Cleaning Solution |
| Packaging | Kit |
| Traceability to NIST | Yes |
| Includes | 475 mL bottle each of pH 4.01, 7.00 and 10.01 buffers; 475 mL bottle of ROSS pH electrode storage solution (810001); 60 mL bottle of pH electrode cleaning solution; and 12 mm diameter electrode storage bottle |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.