Learn More
Thermo Scientific™ Orion™ Potassium Ion Selective Electrodes
Measure potassium ions in aqueous solutions simply, accurately and economically.
Supplier: Thermo Scientific™ 9319BN
Description
Measure potassium ions in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Potassium Electrodes.
- Calibration Standard Catalog number: 0.1M KCl / 921906
- Required ISA Catalog number: 931911
Chemical Identifiers
| drift-free potentials | |
| Sure-Flow Junction | |
| Potassium |
Specifications
| BNC | |
| drift-free potentials | |
| ISE Meter | |
| Potassium |
| Potassium Half Cell Electrode with BNC connector | |
| Sure-Flow Junction | |
| Half Cell Electrode |