missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Electrode Storage Solutions

Restore proper electrode performance and prolong electrode life using these pH electrode storage solutions.

$187.51 - $515.03

Specifications

Type pH Electrode Storage Solution
View More Specs

Products 4
Catalog Number Mfr. No. For Use With (Equipment) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Quantity Price Quantity & Availability  
13303160
View Documents Promotion Details
Thermo Scientific™
810001
ROSS pH Electrode 475 mL
Each for $242.28
Only null left
 
13641908
View Documents Promotion Details
Thermo Scientific™
910001
pH Electrode 475 mL
Each for $187.51
Only null left
 
0935767
View Documents Promotion Details
Thermo Scientific™
910060
pH Electrode 60 mL
Each for $195.67
Only null left
 
1330168
View Documents Promotion Details
Thermo Scientific™
9100CB
pH Electrode 19 L
Each for $515.03
Only null left
 
Description

Description

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Storage Solutions in ready to use bottles.

Storage Solutions:

  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples


Warranty: Out of box

Specifications

Specifications

pH Electrode Storage Solution
Videos
SDS
Documents

Documents

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.