missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Nitrate Ion Selective Electrodes

Measurements are quick, simple, accurate and economical. Analyze free nitrate ions in aqueous solutions reliably at low limits of detection.

$844.71 - $2,952.24

Specifications

Drift drift-free potentials
Junction Sure-Flow Junction
View More Specs

Products 5
Catalog Number Mfr. No. Connector Types For Use With (Equipment) Type Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types For Use With (Equipment) Type Price Quantity & Availability  
13641944
View Documents Promotion Details
Thermo Scientific™
930701
None Nitrate Half-Cell Electrode Replacement Module (3 Pk.)
Pack of 3 Each for $2,135.66
Only null left
 
1330120
View Documents Promotion Details
Thermo Scientific™
930702
None Nitrate Half-Cell Electrode Replacement Module (1 Pk.)
Each for $844.71
Only null left
 
13642261
Accessories View Documents Promotion Details
Thermo Scientific™
9307BNWP
BNC ISE Meter Half Cell Nitrate Electrode
Each for $2,514.12
Only null left
 
1330111
View Documents Promotion Details
Thermo Scientific™
970701
None Combination Nitrate Electrode Replacement Module (1 Pk.)
Pack for $940.90
Only null left
 
13642270
Accessories View Documents Promotion Details
Thermo Scientific™
9707BNWP
BNC ISE Meter Combination Nitrate Electrode
Each for $2,952.24
Only null left
 
Description

Description

Measure nitrate in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Nitrate Electrodes.

The ThermoFisher Orion Nitrate electrode complies with SM 4500-NO3 - D; Orion Method 601 for Drinking water and SM 4500-NO3 - D for Wastewater measurements.

Features:

  • 9707BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. To be used with module 970701.
  • Nitrate Half-Cell Electrode 9307BNWP (comes with half cell module 930702) is to be used with the Orion 900200 sure flow double junction reference electrode.

Solutions required:

  • Standards
    • 920706 0.1 M NaNO3 nitrate calibration standard
    • 920707 1000 ppm as N nitrate calibration standard
    • 930707 100 ppm as N nitrate calibration standard
  • ISA
    • 930711 ISA for nitrate measurements, 475 mL bottle
    • 930710 Nitrate interference suppressor solution (NISS), 475 mL bottle
  • Electrode fill solution
    • 900046 Optimum Results F electrode filling solution, 5 x 60 mL bottles
    • 900002 Inner chamber filling solution for the double junction reference electrode, 5 x 60 mL bottles

Items included with:

  • 9307BNWP
    • 930702 93 series sensing module for 9307BNWP. Quantity 2
    • 930711 ISA for nitrate measurements. 60mL 1 bottle
  • 9707BNWP
    • 970701 97 series sensing module for 9707BNWP. Quantity 2
    • 900046 ISA for nitrate measurements. 60mL 1 bottle

Warranty period:

  • Electrode: 12 months from date of purchase
  • Module: 6 months from date of purchase and 3 months once in operation
Specifications

Specifications

drift-free potentials
Sure-Flow Junction
Videos
SDS
Documents

Documents