missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Calibration Standards

Catalog No. 13641923
Change view
Click to view available options
Analyte or Component Names:
Ammonia
Bromide
Calcium
Carbon Dioxide
Chloride
Chloride and Sodium
Cupric
Fluoride
Iodide
Lead
Low Fluoride
Nitrate
Potassium
Residual Chlorine
Sodium
Sodium Known Addition
Sulfate Standard for Lead ISE
Water Hardness
Concentration:
0.1 M KNO3
0.5 ppm with TISAB II
0.5 ppm with TISAB II and 5 ppm with TISAB II
1 ppm with TISAB II
1 ppm with TISAB II and 10 ppm with TISAB II
10 ppm
10 ppm with TISAB II
100 ppm
1000 ppm
1000 ppm with ISA
2 ppm with TISAB II
0.1 M
0.1M
1M
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Analyte or Component Names Concentration
13641923 Ammonia 0.1M
13641924C Ammonia 1000 ppm
1329998 Ammonia 100 ppm
13641866 Bromide 0.1M
13641811 Calcium 0.1 M
13641768 Carbon Dioxide 0.1 M
13641769 Carbon Dioxide 1000 ppm
13641876 Chloride and Sodium 0.1 M
1329987 Chloride 1000 ppm
13641877 Chloride 100 ppm
13641824 Residual Chlorine 100 ppm
13641835 Cupric 0.1 M
13641871 Fluoride 0.1 M
13641872 Fluoride 100 ppm
13641752 Fluoride 10 ppm with TISAB II
15412 Fluoride 2 ppm with TISAB II
15411 Fluoride 1 ppm with TISAB II
13301140 Fluoride 0.5 ppm with TISAB II
13641879 Fluoride 1 ppm with TISAB II and 10 ppm with TISAB II
13301142 Low Fluoride 0.5 ppm with TISAB II and 5 ppm with TISAB II
13641869 Iodide 0.1 M
13641773 Lead 0.1 M
13641774 Sulfate Standard for Lead ISE 0.1 M
13641888 Nitrate 0.1 M KNO3
13641910 Nitrate 1000 ppm
13641924 Nitrate 100 ppm
13641917 Potassium 0.1 M
13641747 Sodium 1000 ppm
13641909 Sodium 100 ppm
13641948 Sodium 10 ppm
13641787 Sodium Known Addition 1000 ppm with ISA
13641786 Sodium Known Addition 1M
13641862 Water Hardness 100 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
Catalog No. 13641923 Supplier Thermo Scientific™ Supplier No. 951006

Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.

Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.

All standards are 1 pint (475mL) bottles, unless otherwise noted.

Specifications

Description Ammonia Standard
Packaging 1 Bottle
Certificate of Analysis Yes
Concentration or Composition Notes C of A Acceptance Range: 0.1000 M +/- 0.0005 M Ammonium Chloride
Traceability to NIST Yes
Product Line Ammonia Ion Selective Electrodes Including 9512BNWP, 9512HPBNWP
Analyte or Component Names Ammonia
Concentration 0.1M
Color Colorless
For Use With (Equipment) Ammonia Ion Selective Electrode
Solution Type Calibration Standard
Type Calibration Standard
Element Cu
Quantity 475 mL
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.