Learn More
Thermo Scientific™ Orion™ ISE Calibration Standards
Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.
Supplier:  Thermo Scientific™ 948207
 View more versions of this product
Catalog No. 13-641-774
Only null left
Explore available promotions
Description
Description
Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.
All standards are 1 pint (475mL) bottles, unless otherwise noted.
Specifications
Specifications
Sulfate Standard for Lead ISE
Yes
Yes
Liquid
Colorless
Calibration Standard
475 mL
1 Bottle
C of A Acceptance Range: 0.1000 M +/- 0.0005 M Sodium Sulfate
Lead Ion Selective Electrodes Including 9682BNWP
0.1 M NaSO4
Lead Ion Selective Electrode
Calibration Standard
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.