missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Calibration Standards

Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.

Supplier:  Thermo Scientific™ F940914

 View more versions of this product

Catalog No. 13-301-142

Only null left
Explore available promotions

Description

Description

Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.

All standards are 1 pint (475mL) bottles, unless otherwise noted.

Warranty: Out of Box

Specifications

Specifications

Fluoride Low Level Standards
Yes
Yes
Fluoride
0.5 ppm as F- with TISAB II and 5 ppm as F- with TISAB II
Fluoride Ion Selective Electrode
Calibration Standard
8 Bottles (4 Bottles of 0.5 ppm F- Standard Premixed with TISAB II, 4 Bottles of 5 ppm F- Standard Premixed with TISAB II)
C of A Acceptance Range: 0.25 ppm +/- 0.01 ppm Fluoride, 50:50 Premix of Standard to TISAB II (F40904) and 2.50 ppm +/- 0.05 ppm Fluoride, 50:50 Premix of Standard to TISAB II (F40905)
Fluoride Ion Selective Electrodes Including 9609BNWP, 9409BN, 9409SC
Liquid
Colorless
Calibration Standard
475 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.