Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Bromide Combination Ion Selective Electrode
Measure bromide ions in aqueous solutions quickly, simply, accurately and economically.
$2,455.19
Description
Measure bromide ions in aqueous solutions quickly, simply and economically with the Thermo Scientific™ Orion™ Bromide Electrodes.
Features
9635BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
Solutions required
Standard
943506 0.1 M bromide standard, 475 mL bottle
ISA
940011 ISA for bromide measurements, 475 mL bottle
Electrode fill solution
900063 Optimum Results D electrode filling solution, 5 x 60 mL bottles
Items included with
900063 Optimum Results D electrode filling solution, 60 mL 1 bottle
948201 Polishing strip. Qty 1
Warranty period
12 months from date of purchase
Specifications
| Sure-Flow Junction | |
| ISE Meter |