Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Bromide Combination Ion Selective Electrode
Catalog No. 13645158
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Catalog No. Connector Types
13645158 BNC (Waterproof)
1 options
Catalog No. 13645158 Supplier Thermo Scientific™ Supplier No. ORI 9635BNWP
Measure bromide ions in aqueous solutions quickly, simply, accurately and economically.
Measure bromide ions in aqueous solutions quickly, simply and economically with the Thermo Scientific™ Orion™ Bromide Electrodes.
Bromide half-cell electrodes, Part Numbers  9435BN and 9435SC, when used in conjunction with the Thermo Scientific™ Orion™ 900200 double junction reference electrode offers the benefits of the Sure-Flow junction design.
Bromide combination electrode, Part Number 9635BNWP, does not require a separate reference electrode making it convenient to use with small sample volumes.
Related Products
  • Calibration standards: 943506 (0.1 M NaBr)
  • Required ISA: 940011
Specifications
Connector Types BNC (Waterproof)
Description Model 9635; Orion Bromide Combination
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Combination ISE