missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Single Junction Combination pH Electrodes
Confidently measure your samples with the Orion Silver Chloride pH electrodes.
$307.35 - $442.30
Specifications
| pH Range | 0 to 14 |
|---|---|
| For Use With (Equipment) | Russell meters |
| Storage Requirements | Store in Orion pH electrode storage solution 910001 |
| Catalog Number | Mfr. No. | Connector Types | Junction | Type | Material | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Connector Types | Junction | Type | Material | Price | Quantity & Availability | ||
|
13644905
|
Thermo Scientific™
9142BN |
BNC | Ceramic | Combination pH Electrode | Glass Body |
Each for $307.35
|
|
||
|
13644902
|
Thermo Scientific™
9146BN |
BNC | Wick | Combination pH Electrode | Epoxy Body |
Each for $332.18
|
|
||
|
13644903
|
Thermo Scientific™
9147BN |
Requires BNC-to-USS Adapter | HDPE pin | pH/ATC Combination Electrode | Epoxy Body |
Each for $442.30
|
|
||
Description
Thermo Scientific™ Orion™ Silver Chloride pH Electrodes are available in different body materials with either refillable or low maintenance gel options too. The 9142BN is a refillable electrode with chemically resistant glass body best suited for general purpose samples, including solvents, while the 9146BN and 9147BN contain a low-maintenance gel that is best suited for field or plant use, and also educational institutes, where minimal maintenance is preferred. The 9147BN also comes with a built-in ATC.
Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required
Electrode fill solution for 9142BN
900011 4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001 pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
electrode storage solution, 475ml, and 12 mm electrode storage bottle
electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with 9142BN
900011 4M KCl with Ag/AgCl filling solution 60mL 1 bottle
Warranty period
6 months from date of purchase
Specifications
| 0 to 14 | |
| Russell meters | |
| Store in Orion pH electrode storage solution 910001 |