missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS™ Semi-Micro Spear Tip Glass Bodied Combination pH Electrode

Pierce soft and semi-solid samples like cheese, meat, or fruit with a refillable spear tip pH electrode.

$1,776.58 - $1,776.58

Specifications

Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Drift <0.002pH/day
View More Specs

Products 1
Catalog Number Mfr. No. Connector Types Price Quantity  
Catalog Number Mfr. No. Connector Types Price Quantity  
13-644-940
Accessories View Documents
Thermo Scientific™
ORI8163BNWP
BNC N/A
Only null left
 
Description

Description

Thermo Scientific™ Orion™ ROSS Spear Tip Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. Best suited for piercing solid/semi-solid samples and measuring small volume samples.
 

Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
 
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
 
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
 
Warranty period
12 months from date of purchase
Specifications

Specifications

Ceramic
0 to 14
<0.002pH/day
60mL bottle of ROSS electrode fill solution
0.01
32°F to 212°F
20 mm
Glass Body
0°C to 100°C
3M KCl
ROSS
100 μL
Spear
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'