missing translation for 'onlineSavingsMsg'
Learn More

Organophosphorus Pesticides 1 - WP, MilliporeSigma™ Supelco™

Catalog No. 111017353
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017353 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017353 Supplier Merck Emd Millipore Supplier No. QC11612ML

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
CAS 67-64-1
Flash Point -17°C - closed cup - Solvent
Concentration 100% in acetone
Type Certified Reference Material
Quantity 2 mL
Grade Certified Reference Material