missing translation for 'onlineSavingsMsg'
Learn More

Organochlorine Pesticides Ampule 1 - WS, MilliporeSigma™ Supelco™

Catalog No. 111017055
Change view
Click to view available options
Quantity:
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017055 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017055 Supplier Merck Emd Millipore Supplier No. PE15951.2ML
Description This PT sample is packaged as a 1.5 mL concentrate. The solvent for this sample is acetone. The PT sample is not preserved. The sample must be stored at 4C. Sample Preparation PLEASE REFER TO YOUR INSTRUCTION PACKET FOR LATEST INSTRUCTIONS Fill a 1000 mL Class A Volumetric Flask with 900 mL of organic free reagent water. (edit) (delete) Allow the ampule concentrate to equilibrate to room temperature. Open the ampule and pipet exactly 1.0 mL of the concentrate into the flask. Fill to volume and mix well. This represents the sample for analysis. Report results in ug/L

Specifications

CAS 67-64-1
Flash Point -17°C - closed cup - Solvent
Type Proficiency Testing Material
Quantity 1.2 mL