missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Organic phosphorus 3 Monoclonal Antibody (2G11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549667
Description
Antibody tested in indirect ELISA.
Organic phosphorus is an essential nutrient for plants and is commonly found in organic matter, such as decaying plant and animal material. Organic phosphorus 3n is a specific gene target that encodes for a protein involved in the uptake and utilization of organic phosphorus in plants. This gene is important for plant growth and development, as well as for the overall health and productivity of agricultural crops.Specifications
| Organic phosphorus 3 | |
| Monoclonal | |
| 2.6 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of OP-III hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 2G11 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG |