missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Standard, 5000 ppm C, Reagents
Organic Carbon Standard, 5000 ppm C, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CC2052001C
Specifications
| Organic Carbon Standard, 5000 ppm C | |
| 7732-18-5,877-24-7 | |
| Liquid | |
| 4 | |
| Water Analyzer | |
| Water Analysis | |
| Reagent |
| Glass Bottle | |
| 1.005 | |
| Colorless | |
| 100°C | |
| Completely miscible | |
| 1 L |