missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 20 ppm C, Ricca Chemical

Organic Carbon Standard

Supplier:  Ricca Chemical Company R1844200250C

Catalog No. R1844200250


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Organic Carbon Standard, 20 ppm C
877-24-7,7664-38-2,7732-18-5
99.59,0.4,0.0042
Liquid
Colorless
0°C
Miscible with water
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
23676735
Glass Amber Bottle
99.59,0.4,0.0042
1
C8H5KO4
<2
100°C
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
250 mL

Chemical Identifiers

877-24-7
204.222
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 months

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions