missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oregon Green™ 488 Carboxylic Acid, Succinimidyl Ester, 6-isomer

Catalog No. O6149
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O6149 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O6149 Supplier Invitrogen™ Supplier No. O6149

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oregon Green™ 488 Carboxylic Acid, Succinimidyl Ester, 6-isomer

The amine-reactive Oregon Green™ 488 carboxylic acid, succinimidyl ester can be used to can be used to create green fluorescent bioconjugates with excitation/emission maxima ∼496/524 nm. This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∼ 4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.

Specifications

Quantity 5 mg
Product Type Carboxylic Acid Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Oregon Green™ 488
Label Type Classic Dyes
Product Line Oregon Green
Emission 524
Excitation 496
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.