missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Oregon Green™ 488 Azide (Oregon Green™ 6-Carboxamido-(6-Azidohexanyl), Triethylammonium Salt), 6-isomer

Catalog No. O10180
Encompass
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O10180 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O10180 Supplier Molecular Probes™ Supplier No. O10180

Oregon Green™ 488 Azide (Oregon Green™ 6-Carboxamido-(6-Azidohexanyl), Triethylammonium Salt), 6-isomer

The green-fluorescent Oregon Greenr™ 488 azide is reactive with terminal alkynes via a copper-catalyzed click reaction. In addition to being a bright and photostable alternative to fluorescein for use in flow cytometry, microscopy and HCS, Oregon Green™ 488 can also be utilized as a bio-orthogonal or biologically unique hapten for use in applications requiring signal amplification.

Specifications

Format Solid
Quantity 0.5 mg
Product Type Azide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccate and protect from light.
Chemical Reactivity Alkyne
Detection Method Fluorescence
Shipping Condition Room Temperature
System Type Click-iT™
Reactive Moiety Amine, Azide
Label or Dye Oregon Green™ 488
Label Type Classic Dyes
Color Green
Product Line Molecular Probes, Oregon Green
Emission 524
Excitation 496
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.