Learn More
MilliporeSigma™ Supelco™ ORBO™ 77 H2SO4-Treated Carbon Bead (20/30), 500/250 mg W,W,W separators
Catalog No. 111009572
Change view
Click to view available options
Length (Metric):
150 mm
Catalog No. Length (Metric)
111009572 150 mm
1 options
Catalog No. 111009572 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA20036
Used for gas and vapor sampling
ORBO™ 77 H2SO4-Treated Carbon Bead (20/30), 500/250 mg solvent desorption tube is designed for gas and vapor sampling. It contains two beds of the same selective adsorbent separated by glass wool or foam. Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
Specifications
Material W,W,W Separators
Quantity 50/Pk.
Length (Metric) 150 mm
Diameter (Metric) 8 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC