Learn More
MilliporeSigma™ Supelco™ ORBO™ 24 2-HMP on Amberlite XAD-2 (20/40), 150/75 mg W,W,W separators
Catalog No. 111004439
Change view
Click to view available options
Diameter (Metric):
6 mm
Catalog No. Diameter (Metric)
111004439 6 mm
1 options
Catalog No. 111004439 Supplier MilliporeSigma™ Supelco™ Supplier No. 20231
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for gas and vapor sampling
ORBO™ sorbent tubes contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
Specifications
Description Bed A 150 mg/Bed B 75 mg
Material W,W,W Separators
Pore Size 20 to 40 Mesh
Quantity 25/Pk.
Length (Metric) 105 mm
Diameter (Metric) 6 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC