Learn More
MilliporeSigma™ Supelco™ ORBO™ 1000 Precleaned Small PUF Cartridge
Catalog No. 111010444
Change view
Click to view available options
Diameter (Metric):
22 mm
Catalog No. Diameter (Metric)
111010444 22 mm
1 options
Catalog No. 111010444 Supplier MilliporeSigma™ Supelco™ Supplier No. 20557
Used for gas and vapor sampling
For ORBO™ PUF cartridge, several US EPA and ASTM methods require a polyurethane foam (PUF) adsorbent cartridge for monitoring semivolatiles in stack, ambient, indoor, and workplace atmospheres. A low pressure drop across the cartridge facilitates high volume sampling. ORBO-1000 small PUF cartridges are useful for ASTM D4861, EPA IP-8 and EPA TO-10A (Pesticides/PCBs); ASTM D4947 (Chlordane/Heptachlor). Small PUF cartridges are useful for the 1-5 L/min sampling range.
Specifications
Description Assembled
Quantity 3/Pk.
Length (Metric) 7.6 cm
Diameter (Metric) 22 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC