missing translation for 'onlineSavingsMsg'
Learn More

Orange II sodium salt, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014989
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014989 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014989 Supplier Merck Emd Millipore Supplier No. 6914325MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 633-96-5
Physical Form Neat
Linear Formula C16H11N2 NaO4 S
MDL Number MFCD00011657
Synonym 4-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid sodium salt; Acid Orange 7; Acid Orange A; Orange II; Tropaeolin 000 No. 2
Type Analytical Standard
Quantity 25 mg
Formula Weight 350.32
Percent Purity ≥98% (HPLC)
Grade Analytical Standard