missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR6N1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534058
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534058 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534058 Supplier Invitrogen™ Supplier No. PA534058
Only null left

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Hamster, Horse, Rabbit, Platypus (93%).

Olfactory receptors function by associating with odorant molecules to launch a neuronal response that elicits the perception of smell. Olfactory receptor genes are unevenly dispersed among 51 loci on 21 human chromosomes, and are composed of 172 subfamilies. OR6N1 (olfactory receptor, family 6, subfamily N, member 1), also known as olfactory receptor OR1-22, is a 312 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 1 family. As the largest family in the genome, the G-protein coupled receptor 1 family originated from single coding-exon genes. Encoded by a gene that maps to human chromosome 1q23.1, OR6N1 functions as an odorant receptor. Like many hormone and neurotransmitter receptors, OR6N1 shares a 7-transmembrane domain structure and recognizes G protein-mediated transduction of odorant signals.

Specifications

Antigen OR6N1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene OR6N1
Gene Accession No. Q8NGY5
Gene Alias Olfactory receptor 6N1; olfactory receptor family 6 subfamily N member 1; olfactory receptor OR1-22; olfactory receptor, family 6, subfamily N, member 1; OR1-22; OR6N1
Gene Symbols OR6N1
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from 1st extracellular domain of human OR6N1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128372
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less