missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR5V1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103237
Description
Antibody detects endogenous levels of total OR5V1.
Odorant receptor.Chemical Identifiers
| OR5V1 | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| OR5V1 | |
| 6M1-21; hs6M1-21; Olfactory receptor 5V1; olfactory receptor family 5 subfamily V member 1; olfactory receptor OR6-26; olfactory receptor, family 5, subfamily V, member 1; OR5V1 | |
| OR5V1 | |
| Primary | |
| Affinity chromatography |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UGF6 | |
| 81696 | |
| A synthesized peptide derived from human OR5V1(Accession Q9UGF6), corresponding to amino acid residues H244-N294. | |
| Antibody |
Specifications
| OR5V1 | |
| Polyclonal | |
| Unconjugated | |
| OR5V1 | |
| 6M1-21; hs6M1-21; Olfactory receptor 5V1; olfactory receptor family 5 subfamily V member 1; olfactory receptor OR6-26; olfactory receptor, family 5, subfamily V, member 1; OR5V1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 81696 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UGF6 | |
| OR5V1 | |
| A synthesized peptide derived from human OR5V1(Accession Q9UGF6), corresponding to amino acid residues H244-N294. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |