missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR4D6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102194
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102194 Supplier Invitrogen™ Supplier No. PA5102194
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR4D6.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.

Specifications

Antigen OR4D6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR4D6
Gene Accession No. Q8NGJ1
Gene Alias Olfactory receptor 4D6; olfactory receptor family 4 subfamily D member 6; olfactory receptor OR11-250; olfactory receptor, family 4, subfamily D, member 6; OR11-250; OR4D6
Gene Symbols OR4D6
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR4D6(Accession Q8NGJ1), corresponding to amino acid residues M267-G314.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219983
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less