missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR2AG1/OR2AG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa5104388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104388 Supplier Invitrogen™ Supplier No. PA5104388
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR2AG1/2.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.

Specifications

Antigen OR2AG1/OR2AG2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR2AG2
Gene Accession No. A6NM03, Q9H205
Gene Alias HT3; hT3 olfactory receptor; olfactory receptor 2AG1; Olfactory receptor 2AG2; olfactory receptor 2AG3; olfactory receptor family 2 subfamily AG member 1 (gene/pseudogene); olfactory receptor family 2 subfamily AG member 2; olfactory receptor OR11-76 pseudogene; olfactory receptor OR11-79; olfactory receptor, family 2, subfamily AG, member 1; olfactory receptor, family 2, subfamily AG, member 2 pseudogene; olfactory receptor, family 2, subfamily AG, member 3; OR11-76; OR11-79; OR2AG1; OR2AG2; OR2AG2P; OR2AG3
Gene Symbols OR2AG1, OR2AG2
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR2AG1(Accession Q9H205), corresponding to amino acid residues L62-D112.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144125, 338755
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less