missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR1E1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599908 Supplier Invitrogen™ Supplier No. PA599908
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR1E1.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.

Specifications

Antigen OR1E1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR1E1
Gene Accession No. P30953
Gene Alias HGM071; olfactory receptor 13-66; Olfactory receptor 17-2/17-32; olfactory receptor 17-93/17-135/17-136; Olfactory receptor 1E1; olfactory receptor 1E2; Olfactory receptor 1E4; olfactory receptor 1E5; Olfactory receptor 1E6; olfactory receptor 5-85; olfactory receptor family 1 subfamily E member 1; olfactory receptor family 1 subfamily E member 2; olfactory receptor OR17-17; olfactory receptor OR17-18; olfactory receptor OR17-3; olfactory receptor OR17-4; olfactory receptor, family 1, subfamily E, member 1; olfactory receptor, family 1, subfamily E, member 2; olfactory receptor, family 1, subfamily E, member 4; olfactory receptor, family 1, subfamily E, member 5; olfactory receptor, family 1, subfamily E, member 6; olfactory receptor, family 1, subfamily E, member 7; olfactory receptor, family 1, subfamily E, member 8 pseudogene; olfactory receptor, family 1, subfamily E, member 9 pseudogene; olfactory receptor-like protein HGMP07I; OR13-66; OR17-135; OR17-136; OR17-2; OR17-32; OR17-93; OR1E1; OR1E2; OR1E4; OR1E5; OR1E6; OR1E7; OR1E8P; OR1E9P; OR5-85; OST529; OST547
Gene Symbols OR1E1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR1E1(Accession P30953), corresponding to amino acid residues V280-L314.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8387
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less