Learn More
Copan Diagnostics ESwab™Patented Sample Collection and Delivery System
New liquid-based transport for microbiology
$196.84
Specifications
Product Type Specimen Collection Kit
Sterility Sterile
For Use With (Application) Maintains viability of aerobes, anaerobes and fastidious Bacteria for up to 48 Hours
Quantity 50 Pk.
View More Specs
Products 1
Catalog Number Mfr. No. Swab Configuration Price Quantity & Availability  
Catalog Number Mfr. No. Swab Configuration Price Quantity & Availability  
23600900
View Documents Promotion Details
Copan Diagnostics
CA480C
Regular flocked
Pack of 50 Each for $196.84
Case of 10 Pack for $1,968.40
Only null left
Description
Description
New liquid-based transport for microbiology
  • Patented process sprays tips with short strands of nylon fiber that bond in a perpendicular fashion
  • Strong capillary action between the fibers on the flocked tip keeps the sample very close to the surface
  • Sample rapidly and completely elutes from the flocked swab as soon as it is placed in the transport medium
  • Various molded tips correct for different parts of the human anatomy
  • Provides 1mL of sample suspension for Gram stain and multiple culture analysis
  • Maintains viability of aerobes, anaerobes and fastidious bacteria for up to 48 hours
  • Suitable platform for automated liquid handling
Specifications
Specifications
Specimen Collection Kit
Sterile
Maintains viability of aerobes, anaerobes and fastidious Bacteria for up to 48 Hours
50 Pk.