Learn More
Eisco™ Optical Lens
Eisco lenses are spherical, optically worked glass with ground edges and highly polished making them ideal for classroom experiments.
$6.11
Description
- Optical lens
- Choice of double convex or concave
- Choice of diameter and focal length
Specifications
| Each | |
| Spherical | |
| Ground edges, highly polished |