missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Optical Glass Standard Cuvette with Lid

Optical Glass Standard Cuvette features standard rectangular cell with lid for visible/UV spectrophotometry.

Supplier:  Biochrom 80200383

 View more versions of this product

Catalog No. BC80-2003-83


Only null left

Description

Description

  • Optical Glass
  • Standard rectangular cell with lid
  • Fits standard spectrophotometer cell holders
Specifications

Specifications

Type 0 Standard Cuvette
Optical Glass
10 mm
1 mm
46 mm
45 mm
Rectangle
200 μL
12.5 mm
Spacers
190 to 1100 nm
Spectrophotometers, Photometers
Spectrophotometric Measurements
12.7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions