missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OPRD1 Recombinant Rabbit Monoclonal Antibody (SN20-04)
GREENER_CHOICE

Catalog No. PIMA532338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532338 Supplier Invitrogen™ Supplier No. MA532338
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

OPRD1 is an Opioid Receptor that is involved in the control of anxiety, depression, and pain. It is a promising target in the therapy of mood-altering disorders and drug addiction. Delta and kappa opioid receptors function synergistically by forming heterodimers. OPRD1 expression has been reported primarily in the brain. High OPRD1 expression has been documented in the cortex, caudate, hippocampus, nucleus accumbens, and putamen; low OPRD1 expression has been identified in the cerebellum, hypothalamus, substantia nigra, and thalamus. OPRD1 expression is reduced in the putamen and amygdala of Alzheimer's patients and in the putamen in Parkinson's disease.

Specifications

Antigen OPRD1
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone SN20-04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene OPRD1
Gene Accession No. P32300, P33533, P41143
Gene Alias delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a
Gene Symbols OPRD1
Host Species Rabbit
Immunogen Synthetic peptide within Human OPRD1 aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18386, 24613, 4985
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less