Learn More
Applied Biosystems™ OpenArray™ AccuFill™ System Tips
This is a box of tips for use with the OpenArray™ AccuFill™ System, which is part of the OpenArray™ Real-Time PCR Platform.
$185.00 - $1,656.00
Specifications
| Format | Tips |
|---|---|
| For Use With (Equipment) | OpenArray™ |
| Product Line | AccuFill, OpenArray |
| Product Type | System Tip |
| For Use With (Application) | Real Time PCR (qPCR) |
Description
Since AccuFill tips have a shorter shelf-life than OpenArray plates, we recommend that you align your order quantity with your estimated OpenArray throughput.About the OpenArray Real-Time PCR Platform:The OpenArray Real-Time PCR Platform provides PCR-based genotyping and gene expression analysis of hundreds to thousands of samples and assays per day. Researchers can analyze up to 144 samples on a single OpenArray plate. Three OpenArray plates can be cycled and imaged simultaneously on the OpenArray™ Real-Time PCR Instrument, providing high-throughput results with ease. The system workflow thermal cycles and fluorescently images the OpenArray plates. Results are captured on the external PC, and the OpenArray analysis software enables researchers to view specific assay data.
Since AccuFill tips have a shorter shelf-life than OpenArray plates, we recommend that you align your order quantity with your estimated OpenArray throughput.
About the OpenArray Real-Time PCR Platform
The OpenArray Real-Time PCR Platform provides PCR-based genotyping and gene expression analysis of hundreds to thousands of samples and assays per day. Researchers can analyze up to 144 samples on a single OpenArray plate. Three OpenArray plates can be cycled and imaged simultaneously on the OpenArray™ Real-Time PCR Instrument, providing high-throughput results with ease. The system workflow thermal cycles and fluorescently images the OpenArray plates. Results are captured on the external PC, and the OpenArray analysis software enables researchers to view specific assay data.
Complete Platform
The OpenArray Real-Time PCR Platform includes:
• OpenArray Real-Time PCR Instrument—images three OpenArray plates simultaneously
• OpenArray AccuFill System—loads samples into OpenArray plates
• OpenArray™ Case Sealing Station—seals OpenArray plates
• OpenArray instrument accessories (includes PC system)
• OpenArray™ Genotyping Analysis Software
• OpenArray™ Real-Time PCR Analysis Software
Specifications
| Tips | |
| AccuFill, OpenArray | |
| Real Time PCR (qPCR) |
| OpenArray™ | |
| System Tip |
Safety and Handling
ShelfLife : 4 Months from Date of Manufacture
For Research Use Only. Not for use in diagnostic procedures.