Learn More
Thermo Scientific™ Open-Top Screw Caps
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
Supplier: Thermo Scientific™ TS13216
Description
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
- Fit into Reacti-Vial™ small reaction vials
- Used with PTFE/Rubber laminated discs
Chemical Identifiers
| Open Top Screw Cap |
Specifications
| TS-13216 | |
| 15 mm | |
| Open Top Screw Cap |
| 7 mL | |
| 7 mL Screw Cap Septum Vials and Small Reacti-Vials |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.