missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Open-Top Screw Cap

Catalog No. p-7213886
Change view
Click to view available options
Septum:
PTFE/Silicone
PTFE/Silicone/PTFE
No. per Pack:
100
1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum No. per Pack
111008889 PTFE/Silicone/PTFE 1000
111009165 PTFE/Silicone 1000
111011709 PTFE/Silicone 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111008889 Supplier MilliporeSigma™ Supelco™ Supplier No. 27560U
Only null left

Pre-assembled 9 mm polypropylene hole-cap, with septum.

Specifications

Description Clear cap
Septum PTFE/Silicone/PTFE
No. per Pack 1000
Closure Size 9 mm