missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ OPC™ (Oligonucleotide Purification Cartridge), 10/pkg

Catalog No. 400771
Encompass
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400771 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 400771 Supplier Applied Biosystems™ Supplier No. 400771

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains 10 Oligonucleotide Purification cartridges.The Oligonucleotide Purification Cartridge (OPC) cartridge eliminates salts and traces of organic impurities from the crude oligonucleotide, but also provides pure oligonucleotides for sequencing primers, PCR primers, hybridization probes, and gene constructions.

Rapid ResultsThe OPC™ cartridge performs oligonucleotide desalting and purification easily and rapidly. The OPC™ cartridge performs like an affinity column that adsorbs the dimethyltrityl (DMT) group of the trityl-on oligonucleotide while non-DMT bearing failure sequences, by-products, and other impurities wash through. The DMT group is removed with mild acid, allowing the purified, and detritylated oligonucleotide to be eluted.
  • The OPC™ cartridge purifies and deprotects oligonucleotides in one step, saving you time and effort.
  • It eliminates salts, failure sequences, by-products, and trace inorganic impurities to give you oligonucleotides ready for PCR, sequencing, and other applications.
  • It is compatible with our Expedite™ 8909 Synthesizer as well as all ABI synthesizers to offer you flexibility.

Specifications

Column Type Oligonucleotide Synthesis Column
Content And Storage Columns may be stored in a dry environment at room temperature.
Format Column
Sample Type Oligos
For Use With (Equipment) 3900 DNA Synthesizer
For Use With (Application) Oligo Synthesis
No. of Reactions 50 Preps
Product Line OPC
Product Type Oligonucleotide Synthesis Column
Quantity 50 Preps
Shipping Condition Room Temperature
Target Nucleic Acids
Form Solid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.