Learn More
Fisherbrand™ One-Hole Rubber Stoppers
One hole rubber stopper
 View more versions of this product
Catalog No. 14-135F
Only null left
Description
Description
One hole rubber stopper
  • Available in Size No. 00 to 15
  • Hole size: 3 to 5mm
The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).
Specifications
Specifications
Black
4
Inexpensive black natural rubber/SBR blend for general purpose uses
2 cm
Natural Rubber
1
Tapered Stopper (1 hole)
5mm
4
2.6 cm
2.5 cm
0.5cm
Product Suggestions
Product Suggestions