missing translation for 'onlineSavingsMsg'
Learn More

Promega ONE-Glo™ EX Luciferase Assay System

Catalog No. pre8120
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 10 mL
10 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE8120 100 mL
PRE8150 10 x 100 mL
PRE8130 10 x 10 mL
PRE8110 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRE8120 Supplier Promega Supplier No. E8120
Only null left

ONE-Glo™ EX Luciferase Assay System provides both the high sensitivity and long-lived luminescence required to batch-process multiple 96-, 384- or 1,536-well plates. The approximately 2-hour signal half-life provides greater flexibility in assay design.

  • Highly Sensitive Assay with Long-Lived Luminescence
  • Bright, stable signal that can be measured for hours
  • Stable reagent can be stored at room temperature or 4°C
  • Reduced sensitivity to culture media, phenol red and luciferase inhibitors

Specifications

Quantity 100 mL