Learn More
Oncomine™ Tumor Mutation Load Assay, manual library preparation
Specifications
Specifications
| Content And Storage | • 1 Oncomine Tumor Mutation Load Panel, 24 reactions, store at -5 to -30°C. • 1 Ion AmpliSeq Library Kit Plus, 24 reactions, store at -5 to -30°C. |
| Sequencing Type | Genome & DNA Sequencing |
| For Use With (Equipment) | Ion GeneStudio S5 System, Ion S5™ XL System |
| Libraries | Targeted Sequencing Library |
| Product Line | Oncomine |
| Product Type | Tumor Mutation Load Assay |
| Quantity | 24 samples |
| Shipping Condition | Dry Ice |
| Throughput | 24 Samples |
| Workflow Step | Library Generation |
Frequently Asked Questions (FAQs)
No, the Oncomine Tumor Mutation Load Assay is optimized for use with FFPE tissue. The in-house development of the assay specifically focused on three tissue types: melanoma, lung, and colon.
Yes, total variant calls are quantified and reported by the Oncomine Tumor Mutation Load Assay workflow and a .vcf file of all called variants is available to download in the user interface.
Yes, germline filters cover across different ancestry types. They are not ethnicity-based, but are global population-based.
Yes, by following the instructions in the appendix of the User guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0015885_OncomineComprehensiveAssay_v3_UG.pdf).
Yes, Oncomine Comprehensive Assay BAM files can be used with the Oncomine TML workflow. Targets that are not in Oncomine TML workflow will just be discarded during the analysis.