missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OMG Polyclonal Antibody

Description
Keep as concentrated solution. Predicted reactivity: Rhesus Monkey (98%), Bovine (93%). Positive Control: SK-N-SH. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Oligodendrocyte myelin glycoprotein (OMG or OMGP) is a cell adhesion molecule contributing to the interactive process required for myelination in the central nervous system. It is also involved in axonogenesis, neuron projection regeneration, and regulation of collateral sprouting of intact axons in response to injury.
Specifications
Specifications
| Antigen | OMG |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | OMG |
| Gene Accession No. | P23515, Q63912 |
| Gene Alias | oligodendrocyte myelin glycoprotein; oligodendrocyte-myelin glycoprotein; Omg; OMGP |
| Gene Symbols | OMG |
| Show More |