missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole 98.0+%, TCI America™

Catalog No. p-7134294
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03595G 5 g
O035925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03595G Supplier TCI America Supplier No. O03595G
Only null left

Chemical Identifiers

CAS 73590-58-6
Molecular Formula C17H19N3O3S
Molecular Weight (g/mol) 345.417
MDL Number MFCD00083192
InChI Key SUBDBMMJDZJVOS-UHFFFAOYSA-N
Synonym omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
PubChem CID 4594
ChEBI CHEBI:77260
IUPAC Name 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC

Specifications

Melting Point 151°C
Quantity 5 g
Formula Weight 345.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Omeprazole
RTECSNumber DD9087000
TSCA No
Recommended Storage Refrigerator