missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Omeprazole Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 88546-55-8 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C17H19N3O4 S |
| MDL Number | MFCD00869772 |
| Synonym | Omeprazole sulfone; 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulfonyl]-1 H-benzimidazole; 5-Methoxy-2-{[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl}-1 H-benzimidazole; Omeprazole sulphone |
| Type | Certified Reference Material |
| Quantity | 30 mg |
| Formula Weight | 361.42 |
| Grade | Certified Reference Material |