Learn More
Omeprazole, 98%
A proton pump inhibitor | CAS: 73590-58-6 | C17H19N3O3S | 345.417 g/mol
Supplier: Thermo Scientific Chemicals J6286006
Description
Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOmeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.
Solubility
Soluble in water (0.5 mg/ml), DMSO (25 mg/ml), and ethanol (4.5 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications
| Omeprazole | |
| 158°C to 159°C | |
| C17H19N3O3S | |
| 5 g | |
| omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac | |
| SUBDBMMJDZJVOS-UHFFFAOYSA-N | |
| 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole | |
| 4594 | |
| 345.42 | |
| Powder |
| 73590-58-6 | |
| Brown to White | |
| MFCD00083192 | |
| 14,6845 | |
| Soluble in water (0.5mg/ml),DMSO (25mg/ml),and ethanol (4.5mg/ml). | |
| CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC | |
| 345.417 | |
| CHEBI:77260 | |
| 98% |
Chemical Identifiers
| 73590-58-6 | |
| 345.417 | |
| SUBDBMMJDZJVOS-UHFFFAOYSA-N | |
| 4594 | |
| 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole |
| C17H19N3O3S | |
| MFCD00083192 | |
| omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac | |
| CHEBI:77260 | |
| CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : DD9087000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only