missing translation for 'onlineSavingsMsg'
Learn More

omega-Laurinlactam 99.0+%, TCI America™

Catalog No. L009225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L009225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L009225G Supplier TCI America Supplier No. L009225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 947-04-6
Molecular Formula C12H23NO
Molecular Weight (g/mol) 197.32
MDL Number MFCD00005104
InChI Key JHWNWJKBPDFINM-UHFFFAOYSA-N
Synonym Cyclododecanone Isooxime
PubChem CID 13690
IUPAC Name 1-azacyclotridecan-2-one
SMILES O=C1CCCCCCCCCCCN1

Specifications

Melting Point 152°C
Color White
Quantity 25 g
Formula Weight 197.32
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material omega-Laurinlactam
EINECSNumber (5)-1099
RTECSNumber CL6940000
TSCA Yes