accessibility menu, dialog, popup

UserName

CAS RN 947-04-6

IUPAC Name: 1-azacyclotridecan-2-one
Synonyms: nylon-12 laurolactam
Molecular Weight (g/mol): 197.32
Molecular Formula: C12H23NO
InChi Key: JHWNWJKBPDFINM-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCCCCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results